Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Pentadecanesulfonate, TCI America™
SDP

Catalog No. P08891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08891G Supplier TCI America Supplier No. P08891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5896-54-8
Molecular Formula C15H31NaO3S
Molecular Weight (g/mol) 314.46
MDL Number MFCD00059838
InChI Key PLQISZLZPSPBDP-UHFFFAOYSA-M
Synonym 1-Pentadecanesulfonic Acid Sodium Salt
PubChem CID 23670835
IUPAC Name sodium pentadecane-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCCCCS([O-])(=O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 314.46
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Pentadecanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.