CAS RN 59-30-3
Thermo Scientific Chemicals Folic acid, 97%, pure
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.4 MDL Number: MFCD00079305 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
Folic acid, 95-102%
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.404 MDL Number: MFCD00079305 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
Folic acid, ≥95%, Cell Culture Reagent Grade, MP Biomedicals™
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.40 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470
Folic Acid, ≥95%
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.404 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
Folic Acid Hydrate 98.0+%, TCI America™
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.404 MDL Number: MFCD00079305 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
Folic Acid, Powder, USP, 97-102%, Spectrum™ Chemical
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.40 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N IUPAC Name: (2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid SMILES: NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
Folic acid, crystalline
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.404 MDL Number: MFCD00079305 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
Folic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Folic Acid, MP Biomedical
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.40 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470
Medisca Folic Acid, USP (Dietary Supplement Grade)
Folic Acid, USP (Dietary Supplement Grade) is an odorless, crystalline powder that can vary between yellow, yellow-brownish, or yellowish orange in color.
Sigma Aldrich Fmoc-™-2-(7-Octenyl)Ala-Oh
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Folic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Scientific Folic Acid, Fisher BioReagents
CAS: 59-30-3 Molecular Formula: C19H19N7O6 Molecular Weight (g/mol): 441.404 InChI Key: OVBPIULPVIDEAO-LBPRGKRZSA-N Synonym: folic acid,folate,pteroylglutamic acid,vitamin m,folacin,folacid,pteglu,vitamin b9,folvite,pteroyl-l-glutamic acid PubChem CID: 6037 ChEBI: CHEBI:27470 IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N