accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 590-17-0

IUPAC Name: 2-bromoacetonitrile
Synonyms: 2-bromoacetonitrile
Molecular Weight (g/mol): 119.95
Molecular Formula: C2H2BrN
InChi Key: REXUYBKPWIPONM-UHFFFAOYSA-N
SMILES: BrCC#N
Boiling Point 
  • (3)
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)