accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNH2NBr

CAS RN 5900-13-0

IUPAC Name: 5-bromo-3-methoxypyrazin-2-amine
Synonyms: 5-bromo-3-methoxypyrazin-2-amine
Molecular Weight (g/mol): 204.03
Molecular Formula: C5H6BrN3O
InChi Key: QDFJJHGAODFQMN-UHFFFAOYSA-N
SMILES: COC1=NC(Br)=CN=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results