accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONH2N

CAS RN 59084-06-9

IUPAC Name: 4-(2-nitrophenyl)piperazin-1-ium
Synonyms: 4-(2-nitrophenyl)piperazin-1-ium
Molecular Weight (g/mol): 208.24
Molecular Formula: C10H14N3O2
InChi Key: YJRCDSXLKPERNV-UHFFFAOYSA-O
SMILES: [O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results