Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Nitrophenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. N09355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09355G 5 g
N093525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09355G Supplier TCI America Supplier No. N09355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59084-06-9
Molecular Formula C10H14N3O2
Molecular Weight (g/mol) 208.24
MDL Number MFCD00040728
InChI Key YJRCDSXLKPERNV-UHFFFAOYSA-O
Synonym 1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
PubChem CID 100949
IUPAC Name 4-(2-nitrophenyl)piperazin-1-ium
SMILES [O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1

Specifications

Boiling Point 126°C
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Nitrophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.