accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNH2O

CAS RN 591-07-1

IUPAC Name: acetylurea
Synonyms: acetamide, N-(aminocarbonyl)-
Molecular Weight (g/mol): 102.09
Molecular Formula: C3H6N2O2
InChi Key: GKRZNOGGALENQJ-UHFFFAOYSA-N
SMILES: CC(=O)NC(N)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results