Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylurea, 98%, Thermo Scientific™

Catalog No. AAL0834314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0834314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0834314 Supplier Thermo Scientific Chemicals Supplier No. L0834314

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-07-1
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00038140
InChI Key GKRZNOGGALENQJ-UHFFFAOYSA-N
Synonym acetylurea, acetamide, n-aminocarbonyl, n-acetylurea, acetylcarbamide, monoacetylurea, 1-acetylurea, acetic acid, ureide, n-aminocarbonyl acetamide, urea, acetyl, unii-8kru31i919
PubChem CID 68956
IUPAC Name N-carbamoylacetamide
SMILES CC(=O)NC(=O)N

Specifications

Melting Point 214°C to 218°C
Quantity 25 g
Beilstein 1751301
Formula Weight 102.09
Percent Purity 98%
Chemical Name or Material Acetylurea
RTECSNumber AB4331500
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.