accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-08-2

IUPAC Name: acetylthiourea
Synonyms: 1-acetyl-2-thiourea
Molecular Weight (g/mol): 118.15
Molecular Formula: C3H6N2OS
InChi Key: IPCRBOOJBPETMF-UHFFFAOYSA-N
SMILES: CC(=O)NC(N)=S
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)