Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylthiourea 98.0+%, TCI America™
SDP

Catalog No. A011725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A011725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A011725G Supplier TCI America Supplier No. A011725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 591-08-2
Molecular Formula C3H6N2OS
Molecular Weight (g/mol) 118.154
MDL Number MFCD00004937
InChI Key IPCRBOOJBPETMF-UHFFFAOYSA-N
Synonym n-acetylthiourea, acetylthiourea, 1-acetylthiourea, 1-acetyl-2-thiourea, acetothiourea, n-acetyl-2-thiourea, acetamide, n-aminothioxomethyl, n-acetylthiocarbamide, urea, 1-acetyl-2-thio, acetylthiocarbamide
PubChem CID 2723593
IUPAC Name N-carbamothioylacetamide
SMILES CC(=O)NC(=S)N

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 118.15
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylthiourea
RTECSNumber YR7700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.