Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylthiourea, 99+%, Thermo Scientific™

Catalog No. AAB2119836
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2119836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2119836 Supplier Thermo Scientific Chemicals Supplier No. B2119836

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-08-2
Molecular Formula C3H6N2OS
Molecular Weight (g/mol) 118.154
MDL Number MFCD00004937
InChI Key IPCRBOOJBPETMF-UHFFFAOYSA-N
Synonym n-acetylthiourea, acetylthiourea, 1-acetylthiourea, 1-acetyl-2-thiourea, acetothiourea, n-acetyl-2-thiourea, acetamide, n-aminothioxomethyl, n-acetylthiocarbamide, urea, 1-acetyl-2-thio, acetylthiocarbamide
PubChem CID 2723593
IUPAC Name N-carbamothioylacetamide
SMILES CC(=O)NC(=S)N

Specifications

Melting Point 165°C to 169°C
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 969960
Formula Weight 118.16
Percent Purity ≥99%
Chemical Name or Material N-Acetylthiourea
Hazard Category H301
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-699-9
RTECSNumber YR7700000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.