accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59158-14-4

IUPAC Name: methyltrioctylazanium hydrogen sulfate
Synonyms: trioctylmethylammonium hydrogen sulfate
Molecular Weight (g/mol): 465.78
Molecular Formula: C25H55NO4S
InChi Key: MWSPFHZPVVWJCO-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results