Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltri-n-octylammonium Hydrogen Sulfate 97.0+%, TCI America™
SDP

Catalog No. M14575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14575G 5 g
M14571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14575G Supplier TCI America Supplier No. M14575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59158-14-4
Molecular Formula C25H55NO4S
Molecular Weight (g/mol) 465.78
MDL Number MFCD03095545
InChI Key MWSPFHZPVVWJCO-UHFFFAOYSA-M
Synonym Tri-n-octylmethylammonium Hydrogen Sulfate
PubChem CID 10939560
IUPAC Name methyltrioctylazanium hydrogen sulfate
SMILES OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 65°C
Quantity 5 g
Formula Weight 465.78
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyltri-n-octylammonium Hydrogen Sulfate
EINECSNumber (2)-0184
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.