CAS RN 591771-91-4
IUPAC Name:
8-ethyl-4-methyl-2-(2,3,5-trichlorophenyl)-1H,4H,5H,7H,8H-imidazo[2,1-f]purin-5-one hydrochloride
Synonyms:
8-ethyl-4-methyl-2-(2,3,5-trichlorophenyl)-1H,7H,8H-imidazo[2,1-f]purin-5-one hydrochloride
Molecular Weight (g/mol):
435.13
Molecular Formula:
C16H15Cl4N5O
InChi Key:
JUPMOTHIKGWECW-UHFFFAOYNA-N
SMILES:
Cl.CCC1CN2C(=N1)C1=C(N=C(N1)C1=CC(Cl)=CC(Cl)=C1Cl)N(C)C2=O
1
–
1
of
1
results
1
R&D Systems™ PSB 10 hydrochloride
CAS: 591771-91-4 Molecular Formula: C16H15Cl4N5O