accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-05-2

NPbN

CAS RN 592-05-2

IUPAC Name: plumbanedicarbonitrile
Synonyms: plumbanedicarbonitrile
Molecular Weight (g/mol): 259.2
Molecular Formula: C2N2Pb
InChi Key: UMTFPTCPRUEQHM-UHFFFAOYSA-N
SMILES: N#C[Pb]C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results