CAS RN 592474-91-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PKUMDL WQ 2201
Negative allosteric modulator of 3-phosphoglycerate dehydrogenase (PHGDH)