accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOPOOH3COH3C

CAS RN 5927-18-4

IUPAC Name: methyl 2-(dimethoxyphosphoryl)acetate
Synonyms: methyl 2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 182.11
Molecular Formula: C5H11O5P
InChi Key: SIGOIUCRXKUEIG-UHFFFAOYSA-N
SMILES: COC(=O)CP(=O)(OC)OC
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (6)
  • (5)