accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBFFFH3CH3C

CAS RN 593-04-4

IUPAC Name: 1-butoxybutane; trifluoroborane
Synonyms: boron trifluoride; di-N-butyl ether
Molecular Weight (g/mol): 198.04
Molecular Formula: C8H18BF3O
InChi Key: PVKVBDZRILNPJY-UHFFFAOYSA-N
SMILES: FB(F)F.CCCCOCCCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)