Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™

Catalog No. AC428141000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428141000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428141000 Supplier Thermo Scientific Chemicals Supplier No. 428141000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 593-04-4 | C8H18BF3O | 198.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-04-4
Molecular Formula C8H18BF3O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00013195
InChI Key PVKVBDZRILNPJY-UHFFFAOYSA-N
Synonym 1-butoxybutane; trifluoroborane, boron trifluoride dibutyl etherate, boron trifluoride-butyl ether complex, boron, trifluoro 1,1'-oxybis butane-, t-4, boron trifluoride; di-n-butyl ether, 1-butoxybutane-trifluoroborane 1:1, 1-butoxybutane; tris fluoranyl borane, trifluoro 1,1'-oxybis butane boron
PubChem CID 68970
IUPAC Name 1-butoxybutane;trifluoroborane
SMILES FB(F)F.CCCCOCCCC

Specifications

Color Brown to Yellow
Density 0.9590g/mL
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 30-35%
Packaging AcroSeal™ Glass Bottle
Linear Formula BF3O(CH2CH2CH2CH3)2
Quantity 100 mL
Specific Gravity 0.959
Formula Weight 198.03
Physical Form Liquid
Chemical Name or Material Boron trifluoride dibutyl etherate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-783-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.