accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClHONCH2N

CAS RN 5949-11-1

IUPAC Name: hydrogen {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol chloride
Synonyms: hydrogen cinchonidine sulfate chloride
Molecular Weight (g/mol): 330.86
Molecular Formula: C19H23ClN2O
InChi Key: IMUHWLVEEVGMBC-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results