accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59551-75-6

IUPAC Name: 1-bromo-3-(2-methoxyethoxy)propane
Synonyms: 1-bromo-3-(2-methoxyethoxy)propane
Molecular Weight (g/mol): 197.07
Molecular Formula: C6H13BrO2
InChi Key: XXRSNYDVVFLAPC-UHFFFAOYSA-N
SMILES: COCCOCCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results