Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Methoxyethoxy)propyl Bromide 95.0+%, TCI America™
SDP

Catalog No. B18425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18425G 5 g
B18421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18425G Supplier TCI America Supplier No. B18425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59551-75-6
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.072
MDL Number MFCD01321130
InChI Key XXRSNYDVVFLAPC-UHFFFAOYSA-N
Synonym 1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether
PubChem CID 15636133
IUPAC Name 1-bromo-3-(2-methoxyethoxy)propane
SMILES COCCOCCCBr

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 197.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Methoxyethoxy)propyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.