accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOCH3

CAS RN 59576-26-0

IUPAC Name: 1-(4-methylpyridin-2-yl)ethan-1-one
Synonyms: 1-(4-methylpyridin-2-yl)ethanone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: HRVQMQWVGKYDCF-UHFFFAOYSA-N
SMILES: CC(=O)C1=NC=CC(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results