Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A23385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23385G 5 g
A23381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23385G Supplier TCI America Supplier No. A23385G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59576-26-0
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD01863638
InChI Key HRVQMQWVGKYDCF-UHFFFAOYSA-N
Synonym 2-Acetyl-4-picoline
PubChem CID 564760
IUPAC Name 1-(4-methylpyridin-2-yl)ethanone
SMILES CC1=CC(=NC=C1)C(=O)C

Specifications

Melting Point 32°C
Quantity 5 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Acetyl-4-methylpyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.