accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONH2

CAS RN 59578-63-1

IUPAC Name: benzyl(4-hydroxybutyl)azanium
Synonyms: benzyl(4-hydroxybutyl)azanium
Molecular Weight (g/mol): 180.27
Molecular Formula: C11H18NO
InChi Key: SNRCGOVPZJPRQZ-UHFFFAOYSA-O
SMILES: OCCCC[NH2+]CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results