Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylamino-1-butanol 96.0+%, TCI America™
SDP

Catalog No. B15995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15995G 5 g
B159925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15995G Supplier TCI America Supplier No. B15995G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59578-63-1
Molecular Formula C11H18NO
Molecular Weight (g/mol) 180.27
MDL Number MFCD00191609
InChI Key SNRCGOVPZJPRQZ-UHFFFAOYSA-O
Synonym 4-benzylamino-1-butanol, 4-benzylamino butan-1-ol, 4-benzylamino-butan-1-ol, acmc-1ay6n, 4-benzylamino ;butan-1-ol, n-4-hydroxybutyl benzylamine, 1-butanol,4-phenylmethyl amino, 1-butanol, 4-phenylmethyl amino
PubChem CID 1790018
IUPAC Name benzyl(4-hydroxybutyl)azanium
SMILES OCCCC[NH2+]CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 179.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Benzylamino-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.