accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 596-38-3

IUPAC Name: 9-phenyl-9H-xanthen-9-ol
Synonyms: 9-phenylxanthen-9-ol
Molecular Weight (g/mol): 274.32
Molecular Formula: C19H14O2
InChi Key: CVZUPKFPOSRRSK-UHFFFAOYSA-N
SMILES: OC1(C2=CC=CC=C2)C2=CC=CC=C2OC2=CC=CC=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results