accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHONOO

CAS RN 59642-21-6

IUPAC Name: 3-[(4-nitrophenyl)formamido]propanoate
Synonyms: 3-[(4-nitrophenyl)formamido]propanoate
Molecular Weight (g/mol): 237.19
Molecular Formula: C10H9N2O5
InChi Key: PDTLZWITKYGYDN-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)