accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 597-72-8

IUPAC Name: 1-(tripropoxymethoxy)propane
Synonyms: 1-(tripropoxymethoxy)propane
Molecular Weight (g/mol): 248.36
Molecular Formula: C13H28O4
InChi Key: LFMXSZSVDQJYDU-UHFFFAOYSA-N
SMILES: CCCOC(OCCC)(OCCC)OCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results