Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrapropoxymethane 96.0+%, TCI America™
SDP

Catalog No. T12095ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12095ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12095ML Supplier TCI America Supplier No. T12095ML

Chemical Identifiers

CAS 597-72-8
Molecular Formula C13H28O4
Molecular Weight (g/mol) 248.363
MDL Number MFCD00060101
InChI Key LFMXSZSVDQJYDU-UHFFFAOYSA-N
Synonym Tetrapropyl Orthocarbonate
PubChem CID 548455
IUPAC Name 1-(tripropoxymethoxy)propane
SMILES CCCOC(OCCC)(OCCC)OCCC

Specifications

Color Yellow
Boiling Point 224°C
Quantity 5 mL
Formula Weight 248.36
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrapropoxymethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.