accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59733-73-2

IUPAC Name: tetrasodium tris(1,4-dioxane) octakis(methanidylidyneoxidanium) diiron
Synonyms: tetrasodium octakis(carbon monoxide) tris(dioxane) diiron
Molecular Weight (g/mol): 692.05
Molecular Formula: C20H24Fe2Na4O14
InChi Key: GNQHSDPSRBMMNX-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[Na+].[Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1COCCO1.C1COCCO1.C1COCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results