accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 598-30-1

IUPAC Name: (butan-2-yl)lithium
Synonyms: sec-butyllithium
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: VATDYQWILMGLEW-UHFFFAOYNA-N
SMILES: [Li]C(C)CC
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)