Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyllithium, 1.3M in cyclohexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AA43028AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43028AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43028AP Supplier Thermo Scientific Chemicals Supplier No. 43028AP

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-30-1
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009323
InChI Key VATDYQWILMGLEW-UHFFFAOYNA-N
Synonym sec-butyllithium, s-buli, s-butyllithium, s-butyl lithium, unii-5yv3gii1tb, lithium, 1-methylpropyl, 5yv3gii1tb, secbutyllithium, b-butyllithium, sec-butyllitium
PubChem CID 102446
IUPAC Name (butan-2-yl)lithium
SMILES [Li]C(C)CC

Specifications

Color Yellow
Density 0.750 g/mL
Flash Point -17°C (1°F)
Odor Characteristic
Packaging ChemSeal™ bottles
Linear Formula CH3CH2CH(CH3)Li
Quantity 500 mL
UN Number UN3394
Beilstein 3587206
Sensitivity Air and moisture sensitive; May form a precipitate
Formula Weight 64.06
Physical Form Liquid
Chemical Name or Material sec-Butyllithium
Show More Show Less
Hazard Category H225-H250-H260-H304-H314-H335-H336
Hazard Statement GHS H Statement
H224-H250-H260-H252-H304-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 209-927-7
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.