accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59864-04-9

IUPAC Name: 2-[(1Z)-5-fluoro-1-[(4-methanesulfonylphenyl)methylidene]-2-methyl-1H-inden-3-yl]acetic acid
Synonyms: sulindac sulfone exisulind
Molecular Weight (g/mol): 372.41
Molecular Formula: C20H17FO4S
InChi Key: MVGSNCBCUWPVDA-MFOYZWKCSA-N
SMILES: CC1=C(CC(O)=O)C2=CC(F)=CC=C2\C1=C/C1=CC=C(C=C1)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results