Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulindac sulfone, ≥99%, MP Biomedicals™

Catalog No. ICN19603005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19603005 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19603005 Supplier MP Biomedicals, Inc Supplier No. 0219603005

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59864-04-9
Molecular Formula C20H17FO4S
Molecular Weight (g/mol) 372.41
InChI Key MVGSNCBCUWPVDA-RQZCQDPDSA-N
Synonym z-5-fluoro-2-methyl-1-p-methylsulfonyl benzylidene indene-3-acetic acid, sulindac sulfone tlc , solid or powder, 1-4-methylsulfonyl benzylidene-2-methyl-5-fluoro-1h-indene-3-acetic acid, 2-3e-6-fluoro-2-methyl-3-4-methylsulfonylphenyl methylidene inden-1-yl acetic acid
PubChem CID 6158261
IUPAC Name 2-[(3E)-6-fluoro-2-methyl-3-[(4-methylsulfonylphenyl)methylidene]inden-1-yl]acetic acid
SMILES CC1=C(C2=C(C1=CC3=CC=C(C=C3)S(=O)(=O)C)C=CC(=C2)F)CC(=O)O

Specifications

Color Yellow
Assay Percent Range ≥99%
Quantity 5 mg
Formula Weight 372.4
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Sulindac sulfone
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.