CAS RN 59989-18-3
IUPAC Name:
5-ethynyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
5-ethynyluracil
Molecular Weight (g/mol):
136.11
Molecular Formula:
C6H4N2O2
InChi Key:
JOZGNYDSEBIJDH-UHFFFAOYSA-N
SMILES:
O=C1NC=C(C#C)C(=O)N1
1
–
1
of
1
results
1
5-Ethynyluracil 97.0+%, TCI America™
CAS: 59989-18-3 Molecular Formula: C6H4N2O2 Molecular Weight (g/mol): 136.11 MDL Number: MFCD00871973 InChI Key: JOZGNYDSEBIJDH-UHFFFAOYSA-N PubChem CID: 43157 IUPAC Name: 5-ethynyl-1H-pyrimidine-2,4-dione SMILES: C#CC1=CNC(=O)NC1=O