Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethynyluracil 97.0+%, TCI America™
SDP

Catalog No. E1096200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1096200MG 200 mg
E10961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1096200MG Supplier TCI America Supplier No. E1096200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59989-18-3
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD00871973
InChI Key JOZGNYDSEBIJDH-UHFFFAOYSA-N
PubChem CID 43157
IUPAC Name 5-ethynyl-1H-pyrimidine-2,4-dione
SMILES C#CC1=CNC(=O)NC1=O

Specifications

Melting Point 320°C
Quantity 200 mg
Formula Weight 136.11
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethynyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.