CAS RN 60-27-5
Creatinine, MP Biomedicals
CAS: 60-27-5 Molecular Formula: C4H7N3O Molecular Weight (g/mol): 113.12 MDL Number: MFCD00059730 InChI Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N Synonym: creatinine,2-imino-1-methylimidazolidin-4-one,1-methylglycocyamidine,1-methylhydantoin-2-imide,creatinine,heated,4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl,2-amino-1-methylimidazolin-4-one,creatinina,kreatinin,unii-ayi8ex34eu PubChem CID: 588 ChEBI: CHEBI:16737 IUPAC Name: 2-amino-3-methyl-4H-imidazol-5-one SMILES: CN1CC(=O)N=C1N
Creatinine, 98%
CAS: 60-27-5 Molecular Formula: C4H7N3O Molecular Weight (g/mol): 113.12 MDL Number: MFCD00059730 InChI Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N Synonym: creatinine,2-imino-1-methylimidazolidin-4-one,1-methylglycocyamidine,1-methylhydantoin-2-imide,creatinine,heated,4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl,2-amino-1-methylimidazolin-4-one,creatinina,kreatinin,unii-ayi8ex34eu PubChem CID: 588 ChEBI: CHEBI:16737 IUPAC Name: 2-amino-3-methyl-4H-imidazol-5-one SMILES: CN1CC(=O)N=C1N
Creatinine, 98%
CAS: 60-27-5 Molecular Formula: C4H7N3O Molecular Weight (g/mol): 113.12 InChI Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N Synonym: creatinine,2-imino-1-methylimidazolidin-4-one,1-methylglycocyamidine,1-methylhydantoin-2-imide,creatinine,heated,4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl,2-amino-1-methylimidazolin-4-one,creatinina,kreatinin,unii-ayi8ex34eu PubChem CID: 588 ChEBI: CHEBI:16737 IUPAC Name: 2-amino-3-methyl-4H-imidazol-5-one SMILES: CN1CC(=O)N=C1N
Creatinine, NF, 98.5-102%, Spectrum™ Chemical
CAS: 60-27-5 Molecular Formula: C4H7N3O Molecular Weight (g/mol): 113.12 InChI Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N IUPAC Name: 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one SMILES: CN1CC(=O)N=C1N
Creatinine 99.0+%, TCI America™
CAS: 60-27-5 Molecular Formula: C4H7N3O Molecular Weight (g/mol): 113.12 MDL Number: MFCD00059730 InChI Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N Synonym: creatinine,2-imino-1-methylimidazolidin-4-one,1-methylglycocyamidine,1-methylhydantoin-2-imide,creatinine,heated,4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl,2-amino-1-methylimidazolin-4-one,creatinina,kreatinin,unii-ayi8ex34eu PubChem CID: 588 ChEBI: CHEBI:16737 IUPAC Name: 2-amino-3-methyl-4H-imidazol-5-one SMILES: CN1CC(=O)N=C1N
Sigma Aldrich Creatinine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.