Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine 99.0+%, TCI America™
SDP

Catalog No. C0398100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0398100G 100 g
C039825G 25 g
C0398500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0398100G Supplier TCI America Supplier No. C0398100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
MDL Number MFCD00059730
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point 255°C
Color White
Quantity 100 g
Formula Weight 113.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Creatinine
EINECSNumber (9)-0408
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.