Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine, MP Biomedicals

Catalog No. MP210142325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210142325 25 g
MP210142310 10 g
MP210142380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210142325 Supplier MP Biomedicals, Inc Supplier No. 0210142325
Add to Cart
Add to Cart

  • Creatinine is a useful marker for normalizing levels of other molecules found in urine
  • Creatinine is a useful marker for normalizing levels of other molecules found in urine
  • Altered creatinine levels may be associated with conditions that result in decreased renal blood flow, such as diabetes and cardiovascular disease

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
MDL Number MFCD00059730
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point 295°C (literature)
Color White
Linear Formula C4H7N3O
Quantity 25 g
Solubility Information Soluble in 12 parts water; slightly soluble in ethanol; practically insoluble in acetone, ether, chloroform.
Formula Weight 113.1
Percent Purity 98.5 to 102.0%
Grade Anhydrous
Physical Form Powder
Chemical Name or Material CREATININE
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.