CAS RN 600-15-7
IUPAC Name:
2-hydroxybutanoic acid
Synonyms:
α-hydroxybutyric acid (+/-)α-hydoxy butyric acid
Molecular Weight (g/mol):
104.11
Molecular Formula:
C4H8O3
InChi Key:
AFENDNXGAFYKQO-UHFFFAOYNA-N
SMILES:
CCC(O)C(O)=O
1
–
1
of
1
results
1
DL-2-Hydroxybutyric Acid (contains Polymolecular esterification product) 95.0+%, TCI America™
CAS: 600-15-7 Molecular Formula: C4H8O3 Molecular Weight (g/mol): 104.11 MDL Number: MFCD01318568,MFCD00070502 InChI Key: AFENDNXGAFYKQO-UHFFFAOYNA-N PubChem CID: 11266 ChEBI: CHEBI:1148 IUPAC Name: 2-hydroxybutanoic acid SMILES: CCC(O)C(O)=O