accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClCl

CAS RN 601-88-7

IUPAC Name: 1,3-dichloro-2-nitrobenzene
Synonyms: 2,6-dichloro-1-nitrobenzene
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: VITSNECNFNNVQB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results