accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOOHO

CAS RN 6027-21-0

IUPAC Name: 2-amino-4-hydroxybutanoic acid
Synonyms: homoserine Hse
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: UKAUYVFTDYCKQA-UHFFFAOYNA-N
SMILES: NC(CCO)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results