accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N

CAS RN 60272-71-1

IUPAC Name: 2-butyl-6-phenylpyridine
Synonyms: 2-butyl-6-phenylpyridine
Molecular Weight (g/mol): 211.31
Molecular Formula: C15H17N
InChi Key: DSCUZYVYDVDBTP-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results