CAS RN 603-72-5
IUPAC Name:
2λ⁶-thia-3-azatricyclo[6.3.1.0⁴,¹²]dodeca-1(11),4(12),5,7,9-pentaene-2,2-dione
Synonyms:
2λ⁶-thia-3-azatricyclo[6.3.1.0⁴,¹²]dodeca-1(11),4(12),5,7,9-pentaene-2,2-dione
Molecular Weight (g/mol):
205.23
Molecular Formula:
C10H7NO2S
InChi Key:
HOBYPSNNFBRLIX-UHFFFAOYSA-N
SMILES:
O=S1(=O)NC2=C3C1=CC=CC3=CC=C2
1
–
1
of
1
results
1
1,8-Naphthosultam 98%, Thermo Scientific™
CAS: 603-72-5 Molecular Formula: C10H7NO2S Molecular Weight (g/mol): 205.231 InChI Key: HOBYPSNNFBRLIX-UHFFFAOYSA-N Synonym: 1,8-naphthosultam,1,8-naphthalenesultam,2h-naphtho 1,8-cd isothiazole 1,1-dioxide,2h-naphth 1,8-cd isothiazole 1,1-dioxide,2h-naphth 1,8-cd isothiazole,1,1-dioxide,2??-thia-3-azatricyclo 6.3.1.0?, 1 2 dodeca-1 11 ,4 12 ,5,7,9-pentaene-2,2-dione,1,8-naphthasultam,naphtho 1,8-cd isothiazole 1,1-dioxide PubChem CID: 69052 SMILES: C1=CC2=C3C(=C1)NS(=O)(=O)C3=CC=C2