accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SO

CAS RN 6030-36-0

IUPAC Name: 4-methylthiophene-2-carbaldehyde
Synonyms: 4-methylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: HVXCGIPRXBJIRK-UHFFFAOYSA-N
SMILES: CC1=CSC(C=O)=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results