accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCOH3C

CAS RN 60385-06-0

IUPAC Name: 1-isocyanato-4-methoxy-2-methylbenzene
Synonyms: 1-isocyanato-4-methoxy-2-methylbenzene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: XGRQPTXFVDRJMY-UHFFFAOYSA-N
SMILES: COC1=CC=C(N=C=O)C(C)=C1
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results