accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(R)NH2

CAS RN 60419-23-0

IUPAC Name: 1-{[(2R)-pyrrolidin-1-ium-2-yl]methyl}pyrrolidin-1-ium
Synonyms: 1-[(2R)-pyrrolidin-1-ium-2-ylmethyl]pyrrolidin-1-ium
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: YLBWRMSQRFEIEB-SECBINFHSA-P
SMILES: C([C@H]1CCC[NH2+]1)[NH+]1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results