accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3

CAS RN 604785-54-8

IUPAC Name: 9-[4'-(9H-carbazol-9-yl)-2,2'-dimethyl-[1,1'-biphenyl]-4-yl]-9H-carbazole
Synonyms: 9-[4'-(carbazol-9-yl)-2,2'-dimethyl-[1,1'-biphenyl]-4-yl]carbazole
Molecular Weight (g/mol): 512.66
Molecular Formula: C38H28N2
InChi Key: LTUJKAYZIMMJEP-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=C(C)C=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results