accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOONOO

CAS RN 605-69-6

IUPAC Name: 2,4-dinitronaphthalen-1-ol
Synonyms: golden yellow martius yellow
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H6N2O5
InChi Key: FFRBMBIXVSCUFS-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C2=CC=CC=C12)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results